Zirkonium(IV)ethanolat
Lieferant: Apollo Scientific
IN3916-10G
APOSIN3916-10GEA
212
EUR
APOSIN3916-10G
Zirkonium(IV)ethanolat
Zirkonium(IV)ethanolat
Formel:
C₈H₂₀O₄Zr Molecular Weight: 271,47 g/mol Siedepunkt: 235 °C/5mmHg Schmelzpunkt: 171…173 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00015654 CAS-Nummer: 18267-08-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...