Zirconium(IV) isopropoxide isopropanol complex

Lieferant: Apollo Scientific
Gefahr

IN3919-10G IN3919-100G
APOSIN3919-10GEA 157 EUR
APOSIN3919-10G APOSIN3919-100G
Zirconium(IV) isopropoxide isopropanol complex
Zirconium(IV) isopropoxide isopropanol complex
Formel: C₁₅H₃₆O₅Zr
Lagertemperatur: Raumtemperatur
MDL: MFCD00075157
CAS-Nummer: 14717-56-7
UN: 3181
ADR: 4.1,II

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR