5-Amino-o-kresol
Lieferant: Apollo Scientific
|
Achtung
|
OR0011-100G
OR0011-25G
APOSOR0011-100GEA
26.1
EUR
APOSOR0011-100G
APOSOR0011-25G
5-Amino-o-kresol
5-Amino-o-kresol
Formel:
C₇H₉NO Schmelzpunkt: 160…162 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00043922 CAS-Nummer: 2835-95-2 EINECS: 220-618-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...