Thiophen-2-carboxamid
Lieferant: Apollo Scientific
|
Achtung
|
OR0041-25G
APOSOR0041-25GEA
136
EUR
APOSOR0041-25G
Thiophen-2-carboxamid
Thiophen-2-carboxamid
Formel:
C₅H₅NOS Schmelzpunkt: 181…183 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00010425 CAS-Nummer: 5813-89-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...