4-Acetamido-2-methylbenzonitrile

Lieferant: Apollo Scientific
Achtung

Synonyme: 4'-Cyano-3'-methylacetanilide

OR0062-5G
APOSOR0062-5GEA 270 EUR
APOSOR0062-5G
4-Acetamido-2-methylbenzonitrile
4-Acetamido-2-methylbenzonitrile
Formel: C₁₀H₁₀N₂O
Molecular Weight: 174,20 g/mol
Schmelzpunkt: 161…164 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD02258877
CAS-Nummer: 321162-59-8
UN: 3439
ADR: 6.1,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR