4-Nitro-3-picolin-N-oxid 99%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
3-Methyl-4-nitropyridin-1-oxid, 4-Nitro-3-picolin-N-oxid, 3-Methyl-4-nitropyridin-N-oxid
OR0074-5G
APOSOR0074-5GEA
21.9
EUR
APOSOR0074-5G
4-Nitro-3-picolin-N-oxid 99%
4-Nitro-3-picolin-N-oxid
Formel:
C₆H₆N₂O₃ Molecular Weight: 154,13 g/mol Schmelzpunkt: 134…138 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00014626 CAS-Nummer: 1074-98-2 EINECS: 214-050-8 UN: 2811 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...