Chinoxalin-6-amin

Lieferant: Apollo Scientific

Synonyme: 6-Aminoquinoxaline

OR0088-1G
APOSOR0088-1GEA 0 EUR
APOSOR0088-1G
Chinoxalin-6-amin
Chinoxalin-6-amin
Formel: C₈H₇N₃
Molecular Weight: 145,1600 g/mol
MDL: MFCD00462821
CAS-Nummer: 6298-37-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR