2-Benzo[b]thiophenboronsäure
Lieferant: Apollo Scientific
|
Achtung
|
OR0093-1G
OR0093-25G
OR0093-5G
APOSOR0093-1GEA
21.9
EUR
APOSOR0093-1G
APOSOR0093-25G
APOSOR0093-5G
2-Benzo[b]thiophenboronsäure
2-Benzo[b]thiophenboronsäure
Formel:
C₈H₇BO₂S Schmelzpunkt: 252…255 °C Lagertemperatur: Gefrierschrank |
MDL:
MFCD01075674 CAS-Nummer: 98437-23-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...