3-Chlor-2-methylbenzolsulfonamid

Lieferant: Apollo Scientific
Achtung

OR0114-250MG
APOSOR0114-250MGEA 0 EUR
APOSOR0114-250MG
3-Chlor-2-methylbenzolsulfonamid
3-Chlor-2-methylbenzolsulfonamid
Formel: C₇H₈ClNO₂S
Lagertemperatur: Raumtemperatur
MDL: MFCD00234295
CAS-Nummer: 3476-03-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR