4-Chlorophenyl cyanomethyl sulphone 98%

Lieferant: Apollo Scientific
Achtung

Synonyme: 4-Chlorophenylsulphonylacetonitrile, 2-((4-Chlorophenyl)sulphonyl)acetonitrile

OR0123-5G
APOSOR0123-5GEA 125 EUR
APOSOR0123-5G
4-Chlorophenyl cyanomethyl sulphone 98%
4-Chlorophenyl cyanomethyl sulphone
Formel: C₈H₆ClNO₂S
Molecular Weight: 215,65 g/mol
Schmelzpunkt: 168…170 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00045626
CAS-Nummer: 1851-09-8
UN: 3439
ADR: 6.1,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR