4-Methyl-5-(3-thienylmethyl)-4H-1,2,4-triazole-3-thiol
Lieferant: Apollo Scientific
|
Gefahr
|
Synonyme:
4-Methyl-3-[(thien-3-yl)methyl]-5-thio-4H-1,2,4-triazole
OR0143-1G
APOSOR0143-1GEA
92.8
EUR
APOSOR0143-1G
4-Methyl-5-(3-thienylmethyl)-4H-1,2,4-triazole-3-thiol
4-Methyl-5-(3-thienylmethyl)-4H-1,2,4-triazole-3-thiol
Formel:
C₈H₉N₃S₂ Molecular Weight: 211,31 g/mol Schmelzpunkt: 130…133 °C Lagertemperatur: Kühlschrank |
MDL:
MFCD00832943 CAS-Nummer: 262610-50-4 UN: 1325 ADR: 4.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...