Methyl-2,4-dioxo-4-pyridin-3-ylbutanoate 95%

Lieferant: Apollo Scientific
Achtung

OR01483-500MG
APOSOR01483-500MGEA 58 EUR
APOSOR01483-500MG
Methyl-2,4-dioxo-4-pyridin-3-ylbutanoate 95%
Methyl-2,4-dioxo-4-pyridin-3-ylbutanoate
Formel: C₁₀H₉NO₄
Molecular Weight: 207,19 g/mol
Schmelzpunkt: 119 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD06245328
CAS-Nummer: 23424-36-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR