2-[(2-Aminoethyl)amino]nicotinic acid

Lieferant: Apollo Scientific
Achtung

Synonyme: 2-[(2-Aminoethyl)amino]-3-carboxypyridine, 2-[(2-Aminoethyl)amino]pyridine-3-carboxylic acid

OR01488-250MG
APOSOR01488-250MGEA 279 EUR
APOSOR01488-250MG
2-[(2-Aminoethyl)amino]nicotinic acid
2-[(2-Aminoethyl)amino]nicotinic acid
Formel: C₈H₁₁N₃O₂
Molecular Weight: 181,192 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD06245331
CAS-Nummer: 374063-93-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR