4-Chlor-2,5-dimethoxyphenylisothiocyanat
Lieferant: Apollo Scientific
|
Achtung
|
OR014888-1G
APOSOR014888-1GEA
30.6
EUR
APOSOR014888-1G
4-Chlor-2,5-dimethoxyphenylisothiocyanat
4-Chlor-2,5-dimethoxyphenylisothiocyanat
Formel:
C₉H₈ClNO₂S Schmelzpunkt: 87…88 °C Lagertemperatur: Kühlschrank |
MDL:
MFCD00046809 CAS-Nummer: 306935-82-0 UN: 2811 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...