5-Chinoxalincarbonsäure

Lieferant: Apollo Scientific
Achtung

Synonyme: Quinoxaline-5-carboxylic acid

OR01489-100MG
APOSOR01489-100MGEA 30.6 EUR
APOSOR01489-100MG
5-Chinoxalincarbonsäure
5-Quinoxalinecarboxylic acid
Formel: C₉H₆N₂O₂
Molecular Weight: 174,1561 g/mol
Schmelzpunkt: 167…172 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD02854494
CAS-Nummer: 6924-66-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR