7-Bromochroman-4-one

Lieferant: Apollo Scientific

OR015010-5G
APOSOR015010-5GEA 63.8 EUR
APOSOR015010-5G
7-Bromochroman-4-one
7-Bromochroman-4-one
Formel: C₉H₇BrO₂
Lagertemperatur: Raumtemperatur
MDL: MFCD09744055
CAS-Nummer: 18442-22-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR