5-Pyridin-2-yl-1H-pyrazole-3-carboxylic acid 97%
Lieferant: Apollo Scientific
|
Achtung
|
OR01503-500MG
APOSOR01503-500MGEA
21.9
EUR
APOSOR01503-500MG
5-Pyridin-2-yl-1H-pyrazole-3-carboxylic acid 97%
5-Pyridin-2-yl-1H-pyrazole-3-carboxylic acid
Formel:
C9H7N3O2 Schmelzpunkt: 300 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD05678901 CAS-Nummer: 374064-02-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...