Pyrimidine-4-carbonitrile

Lieferant: Apollo Scientific

Synonyme: 4-Cyanopyrimidine

OR015037-1G
APOSOR015037-1GEA 227 EUR
APOSOR015037-1G
Pyrimidine-4-carbonitrile
Pyrimidine-4-carbonitrile
Formel: C5H3N3
Schmelzpunkt: 30…31 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD07787613
CAS-Nummer: 42839-04-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR