N-(4-Aminophenyl)-N-methyl-2-(4-methylpiperazin-1-yl)acetamide

Lieferant: Apollo Scientific

OR015042-1G
APOSOR015042-1GEA 21.5 EUR
APOSOR015042-1G
N-(4-Aminophenyl)-N-methyl-2-(4-methylpiperazin-1-yl)acetamide
N-(4-Aminophenyl)-N-methyl-2-(4-methylpiperazin-1-yl)acetamide
Formel: C₁₄H₂₂N₄O
Lagertemperatur: Raumtemperatur
MDL: MFCD12457658
CAS-Nummer: 262368-30-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR