3-[4-(Morpholin-4-yl)-3-nitrophenyl]acrylic acid

Lieferant: Apollo Scientific
Achtung

Synonyme: 4-(Morpholin-4-yl)-3-nitrocinnamic acid, 3-[4-(Morpholin-4-yl)-3-nitrophenyl]prop-2-enoic acid

OR015097-250MG
APOSOR015097-250MGEA 21.9 EUR
APOSOR015097-250MG
3-[4-(Morpholin-4-yl)-3-nitrophenyl]acrylic acid
3-[4-(Morpholin-4-yl)-3-nitrophenyl]acrylic acid
Formel: C₁₃H₁₄N₂O₅
Molecular Weight: 278,261 g/mol
Schmelzpunkt: 185…187 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00831764
CAS-Nummer: 300541-93-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR