2,4-Dioxo-4-(pyridin-3-yl)butanoic acid 95%

Lieferant: Apollo Scientific
Achtung

OR01522-1G
APOSOR01522-1GEA 509 EUR
APOSOR01522-1G
2,4-Dioxo-4-(pyridin-3-yl)butanoic acid 95%
2,4-Dioxo-4-(pyridin-3-yl)butanoic acid
Formel: C₉H₇NO₄
Molecular Weight: 193,156 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD06245355
CAS-Nummer: 394655-14-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR