N'-Hydroxy-4-[(2-methyl-1,3-thiazol-4-yl)methoxy]benzenecarboximidamide 95%

Lieferant: Apollo Scientific
Achtung

Synonyme: 4-(2-Methylthiazol-4-ylmethoxy)benzamidoxime

OR015254-1G
APOSOR015254-1GEA 116 EUR
APOSOR015254-1G
N'-Hydroxy-4-[(2-methyl-1,3-thiazol-4-yl)methoxy]benzenecarboximidamide 95%
N'-Hydroxy-4-[(2-methyl-1,3-thiazol-4-yl)methoxy]benzenecarboximidamide
Formel: C₁₂H₁₃N₃O₂S
Molecular Weight: 263,316 g/mol
Schmelzpunkt: 190…192 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00830779
CAS-Nummer: 306935-19-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR