1-(4-Methoxyphenyl)-2-(quinoxalin-2-yl)ethanol 95+%

Lieferant: Apollo Scientific

OR0154-10G OR0154-1G OR0154-5G
APOSOR0154-10GEA 606 EUR
APOSOR0154-10G APOSOR0154-1G APOSOR0154-5G
1-(4-Methoxyphenyl)-2-(quinoxalin-2-yl)ethanol 95+%
1-(4-Methoxyphenyl)-2-(quinoxalin-2-yl)ethanol
Formel: C₁₇H₁₆N₂O₂
Lagertemperatur: Raumtemperatur
MDL: MFCD03426056
CAS-Nummer: 849021-36-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR