3-Bromoimidazo[1,2-a]pyrazine
Lieferant: Apollo Scientific
|
Gefahr
|
OR01540-5G
APOSOR01540-5GEA
114
EUR
APOSOR01540-5G
3-Bromoimidazo[1,2-a]pyrazine
3-Bromoimidazo[1,2-a]pyrazine
Formel:
C6H4BrN3 Schmelzpunkt: 191…196 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD06245372 CAS-Nummer: 57948-41-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...