5-Methyl-3-(4-pyridyl)-1,2,4-oxadiazole 97%

Lieferant: Apollo Scientific
Achtung

Synonyme: 4-(5-Methyl-1,2,4-oxadiazol-3-yl)pyridine

OR01543-50MG
APOSOR01543-50MGEA 21.9 EUR
APOSOR01543-50MG
5-Methyl-3-(4-pyridyl)-1,2,4-oxadiazole 97%
5-Methyl-3-(4-pyridyl)-1,2,4-oxadiazole
Formel: C₈H₇N₃O
Molecular Weight: 161.16 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00464110
CAS-Nummer: 10350-70-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR