N-[(Benzyloxy)carbonyl]-L-valin

Lieferant: Apollo Scientific
Achtung

Synonyme: N-Cbz-L-valine, Z-L-Val-OH, Z-Val-OH

OR01545-5G
APOSOR01545-5GEA 21.9 EUR
APOSOR01545-5G
N-[(Benzyloxy)carbonyl]-L-valin
N-Carbobenzyloxy-L-valine
Formel: C₁₃H₁₇NO₄
Molecular Weight: 251,3 g/mol
Schmelzpunkt: 59…60 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00008922
CAS-Nummer: 1149-26-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR