3-Nitroimidazo[1,2-a]pyridin 95%
Lieferant: Apollo Scientific
|
Achtung
|
OR01548-1G
APOSOR01548-1GEA
285
EUR
APOSOR01548-1G
3-Nitroimidazo[1,2-a]pyridin 95%
3-Nitroimidazo[1,2-a]pyridin
Formel:
C₇H₅N₃O₂ Schmelzpunkt: 204…206 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00014557 CAS-Nummer: 4926-45-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...