1-(2-Pyridyl)ethylamine

Lieferant: Apollo Scientific
Gefahr

OR01560-5G OR01560-25G
APOSOR01560-5GEA 0 EUR
APOSOR01560-5G APOSOR01560-25G
1-(2-Pyridyl)ethylamine
1-(2-Pyridyl)ethylamine
Formel: C₇H₁₀N₂
Lagertemperatur: Raumtemperatur
MDL: MFCD05215238
CAS-Nummer: 42088-91-5
UN: 2735
ADR: 8,III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR