4-(1-Aminoethyl)pyridin 97%
Lieferant: Apollo Scientific
|
Gefahr
|
Synonyme:
1-(4-Pyridyl)ethylamin, 1-Pyridin-4-ylethylamin
OR01563-1G
APOSOR01563-1GEA
27.7
EUR
APOSOR01563-1G
4-(1-Aminoethyl)pyridin 97%
4-(1-Aminoethyl)pyridine
Formel:
C₇H₁₀N₂ Molecular Weight: 122,17 g/mol Dichte: 1,019 g/cm³ (25 °C) Flammpunkt: 104,4 °C Lagertemperatur: Kühlschrank |
MDL:
MFCD02245004 CAS-Nummer: 50392-78-4 UN: 2922 ADR: 8+6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...