2-(2-Pyridyl)indole

Lieferant: Apollo Scientific
Achtung

Synonyme: 2-(Pyridin-2-yl)-1H-indole

OR01564-1G
APOSOR01564-1GEA 194 EUR
APOSOR01564-1G
2-(2-Pyridyl)indole
2-(2-Pyridyl)indole
Formel: C₁₃H₁₀N₂
Molecular Weight: 194,24 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00033466
CAS-Nummer: 13228-40-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR