1-[5-(4-Chlorophenyl)-2-methylfur-3-yl]ethan-1-one
Lieferant: Apollo Scientific
|
Achtung
|
OR01574-1G
APOSOR01574-1GEA
92.8
EUR
APOSOR01574-1G
1-[5-(4-Chlorophenyl)-2-methylfur-3-yl]ethan-1-one
1-[5-(4-Chlorophenyl)-2-methylfur-3-yl]ethan-1-one
Formel:
C₁₃H₁₁ClO₂ Schmelzpunkt: 114…115 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00832873 CAS-Nummer: 43020-12-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...