2,3-Dihydro-4-(3-hydroxyphenyl)-1H-1,5-benzodiazepine

Lieferant: Apollo Scientific

Synonyme: 3-(4,5-Dihydro-3H-1,4-benzodiazepin-2-yl)phenol

OR01589-50MG
APOSOR01589-50MGEA 400 EUR
APOSOR01589-50MG
2,3-Dihydro-4-(3-hydroxyphenyl)-1H-1,5-benzodiazepine
2,3-Dihydro-4-(3-hydroxyphenyl)-1H-1,5-benzodiazepine
Formel: C₁₅H₁₄N₂O
Lagertemperatur: Raumtemperatur
MDL: MFCD06245402
CAS-Nummer: 889955-12-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR