4'-Brom-2'-chloracetanilid

Lieferant: Apollo Scientific
Achtung

Synonyme: N-(4-Brom-2-chlorphenyl)acetamid

OR015905-25G
APOSOR015905-25GEA 89.9 EUR
APOSOR015905-25G
4'-Brom-2'-chloracetanilid
4'-Brom-2'-chloracetanilid
Formel: H₃CC(O)NH(C₆H₃BrCl)
Molecular Weight: 248,51 g/mol
Schmelzpunkt: 158…160 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00040852
CAS-Nummer: 3460-23-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR