4-Chloro-1H-pyrazole-3-carboxaldehyde

Lieferant: Apollo Scientific
Achtung

Synonyme: 4-Chloro-1H-pyrazole-5-carbaldehyde, 4-Chloro-3-formylpyrazole

OR01601-1G
APOSOR01601-1GEA 348 EUR
APOSOR01601-1G
4-Chloro-1H-pyrazole-3-carboxaldehyde
4-Chloro-1H-pyrazole-3-carboxaldehyde
Formel: C₄H₃ClN₂O
Molecular Weight: 130,53 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00110641
CAS-Nummer: 623570-54-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR