2-Bromo-1-(4-isobutylphenyl)propan-1-one
Lieferant: Apollo Scientific
|
Gefahr
|
OR01605-1G
APOSOR01605-1GEA
0
EUR
APOSOR01605-1G
2-Bromo-1-(4-isobutylphenyl)propan-1-one
2-Bromo-1-(4-isobutylphenyl)propan-1-one
Formel:
C₁₃H₁₇BrO Schmelzpunkt: 60…62 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00831700 CAS-Nummer: 80336-64-7 UN: 3261 ADR: 8,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...