2-Amino-4-methylthiophene-3-carbonitrile

Lieferant: Apollo Scientific
Achtung

Synonyme: 2-Amino-3-cyano-4-methylthiophene

OR01621-1G
APOSOR01621-1GEA 30.6 EUR
APOSOR01621-1G
2-Amino-4-methylthiophene-3-carbonitrile
2-Amino-4-methylthiophene-3-carbonitrile
Formel: C₆H₆N₂S
Molecular Weight: 138,19 g/mol
Schmelzpunkt: 119…120 °C
Lagertemperatur: Kühlschrank
MDL: MFCD00277568
CAS-Nummer: 4623-55-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR