Ethyl 1-naphthoylformate 97%

Lieferant: Apollo Scientific
Achtung

Synonyme: Ethyl 2-(naphthalen-1-yl)-2-oxoacetate, Ethyl (naphth-1-yl)(oxo)acetate

OR016324-1G
APOSOR016324-1GEA 522 EUR
APOSOR016324-1G
Ethyl 1-naphthoylformate 97%
Ethyl-1-naphthoylformate
Formel: C₁₄H₁₂O₃
Molecular Weight: 228.25 g/mol
Siedepunkt: 152…154 °C/0.2mmHg
Lagertemperatur: Raumtemperatur
MDL: MFCD01319637
CAS-Nummer: 33656-65-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR