4-[(Pyridin-3-ylmethyl)amino]piperidine, N1-BOC protected 95+%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
tert-Butyl 4-[(pyridin-3-ylmethyl)amino]piperidine-1-carboxylate
OR01643-1G
APOSOR01643-1GEA
122
EUR
APOSOR01643-1G
4-[(Pyridin-3-ylmethyl)amino]piperidine, N1-BOC protected 95+%
4-[(Pyridin-3-ylmethyl)amino]piperidine, N1-BOC protected
Formel:
C₁₆H₂₅N₃O₂ Schmelzpunkt: 45…48 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD04114984 CAS-Nummer: 206274-21-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...