tert-Butyl-4-aminophenylacetate 95+%

Lieferant: Apollo Scientific
Achtung

Synonyme: tert-Butyl 2-(4-aminophenyl)acetate

OR01647-5G
APOSOR01647-5GEA 140 EUR
APOSOR01647-5G
tert-Butyl-4-aminophenylacetate 95+%
tert-Butyl-4-aminophenylacetate
Formel: C₁₂H₁₇NO₂
Molecular Weight: 207.27 g/mol
Schmelzpunkt: 30…32 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD06245494
CAS-Nummer: 174579-31-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR