1-Amino-3-(4-methoxyphenoxy)propan-2-ol
Lieferant: Apollo Scientific
|
Gefahr
|
Synonyme:
4-(3-Amino-2-hydroxypropoxy)anisole
OR016549-1G
APOSOR016549-1GEA
116
EUR
APOSOR016549-1G
1-Amino-3-(4-methoxyphenoxy)propan-2-ol
1-Amino-3-(4-methoxyphenoxy)propan-2-ol
Formel:
C₁₀H₁₅NO₃ Molecular Weight: 197,24 g/mol Schmelzpunkt: 110…112 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00205693 CAS-Nummer: 5002-93-7 UN: 3259 ADR: 8,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...