N-(1-Benzylpiperidin-4-yl)benzene-1,2-diamine, tech

Lieferant: Apollo Scientific
Gefahr

OR01657-250MG
APOSOR01657-250MGEA 79.9 EUR
APOSOR01657-250MG
N-(1-Benzylpiperidin-4-yl)benzene-1,2-diamine, tech
N-(1-Benzylpiperidin-4-yl)benzene-1,2-diamine
Formel: C₁₈H₂₃N₃
Molecular Weight: 281,395 g/mol
Schmelzpunkt: 111…112 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00023740
CAS-Nummer: 57718-47-5
EINECS: 260-916-3
UN: 3259
ADR: 8,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR