5-(tert-Butyl)thiophene-2-carboxylic acid

Lieferant: Apollo Scientific
Gefahr

OR01670-1G OR01670-5G
APOSOR01670-1GEA 45.1 EUR
APOSOR01670-1G APOSOR01670-5G
5-(tert-Butyl)thiophene-2-carboxylic acid
5-(tert-Butyl)thiophene-2-carboxylic acid
Formel: C₉H₁₂O₂S
Schmelzpunkt: 125…127 °C
Lagertemperatur: Kühlschrank
MDL: MFCD01566583
CAS-Nummer: 29212-25-7
UN: 1759
ADR: 8,III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR