3-(Thien-2-yl)piperazine, N1-BOC protected

Lieferant: Apollo Scientific
Achtung

Synonyme: tert-Butyl 3-(thien-2-yl)piperazine-1-carboxylate, 1-(tert-Butoxycarbonyl)-3-(thien-2-yl)piperazine

OR01674-50MG
APOSOR01674-50MGEA 58 EUR
APOSOR01674-50MG
3-(Thien-2-yl)piperazine, N1-BOC protected
3-(Thien-2-yl)piperazine, N1-BOC protected
Formel: C₁₃H₂₀N₂O₂S
Lagertemperatur: Kühlschrank
MDL: MFCD06660372
CAS-Nummer: 886771-38-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR