3-Phenylquinoxaline-5-carboxylic acid
Lieferant: Apollo Scientific
|
Achtung
|
OR01681-100MG
APOSOR01681-100MGEA
58
EUR
APOSOR01681-100MG
3-Phenylquinoxaline-5-carboxylic acid
3-Phenylquinoxaline-5-carboxylic acid
Formel:
C₁₅H₁₀N₂O₂ Molecular Weight: 250.252 g/mol |
MDL:
MFCD06660374 CAS-Nummer: 162135-93-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...