9-Amino-1,2,3,4-tetrahydro-5H-1,4-benzodiazepin-5-one 95%

Lieferant: Apollo Scientific

Synonyme: 9-Amino-1,2,3,4-tetrahydro-1,4-benzo[e]diazepin-5-one

OR01685-1G
APOSOR01685-1GEA 723 EUR
APOSOR01685-1G
9-Amino-1,2,3,4-tetrahydro-5H-1,4-benzodiazepin-5-one 95%
9-Amino-1,2,3,4-tetrahydro-5H-1,4-benzodiazepin-5-one
Formel: C₉H₁₁N₃O
Molecular Weight: 177,203 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD03265458
CAS-Nummer: 328546-66-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR