2-(4-Methoxyphenyl)piperazine

Lieferant: Apollo Scientific
Gefahr

OR01693-50MG
APOSOR01693-50MGEA 69.6 EUR
APOSOR01693-50MG
2-(4-Methoxyphenyl)piperazine
2-(4-Methoxyphenyl)piperazine
Formel: C₁₁H₁₆N₂O
Molecular Weight: 192,26 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD05864677
CAS-Nummer: 91517-26-9
UN: 3259
ADR: 8,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR