1-(4-Biphenylyl)ethylamine
Lieferant: Apollo Scientific
|
Achtung
|
OR01696-250MG
APOSOR01696-250MGEA
0
EUR
APOSOR01696-250MG
1-(4-Biphenylyl)ethylamine
1-(4-Biphenylyl)ethylamine
Formel:
C₁₄H₁₅N Lagertemperatur: Raumtemperatur |
MDL:
MFCD02667819 CAS-Nummer: 86217-82-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...