4-Aminobenzaldehyde, N-BOC protected 97%

Lieferant: Apollo Scientific
Gefahr

Synonyme: (4-Formyl-phenyl)-carbamicacidtert-butylester, tert-Butyl (4-formylphenyl)carbamate

OR01705-5G
APOSOR01705-5GEA 156 EUR
APOSOR01705-5G
4-Aminobenzaldehyde, N-BOC protected 97%
4-Aminobenzaldehyde, N-BOC protected
Formel: C₁₂H₁₅NO₃
Molecular Weight: 221.252 g/mol
Schmelzpunkt: 138…142 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD06245541
CAS-Nummer: 144072-30-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR