6-[(2-Aminoethyl)amino]nicotinic acid

Lieferant: Apollo Scientific
Achtung

Synonyme: 6-[(2-Aminoethyl)amino]pyridine-3-carboxylic acid, 2-[(2-Aminoethyl)amino]-5-carboxypyridine

OR01708-250MG
APOSOR01708-250MGEA 140 EUR
APOSOR01708-250MG
6-[(2-Aminoethyl)amino]nicotinic acid
6-[(2-Aminoethyl)amino]nicotinic acid
Formel: C₈H₁₁N₃O₂
Molecular Weight: 181,192 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD06245441
CAS-Nummer: 904815-02-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR