6-({2-[(tert-Butoxycarbonyl)amino]ethyl}amino)nicotinic acid
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
6-({2-[(tert-Butoxycarbonyl)amino]ethyl}amino)pyridine-3-carboxylic acid, 2-({2-[(tert-Butoxycarbonyl)amino]ethyl}amino)-5-carboxypyridine
OR01709-50MG
APOSOR01709-50MGEA
69.6
EUR
APOSOR01709-50MG
6-({2-[(tert-Butoxycarbonyl)amino]ethyl}amino)nicotinic acid
6-({2-[(tert-Butoxycarbonyl)amino]ethyl}amino)nicotinic acid
Formel:
C₁₃H₁₉N₃O₄ Molecular Weight: 281.308 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD06245544 CAS-Nummer: 904815-08-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...